Bottles, wide neck, round, with screw cap
Katalognr. 215-3585
Supplier: PONT PACKAGING
Specifications
- Filettatura:38/R3
- Capacità:500 ml
- Seq:02
- Conf:33
Specifications
About this item
Glass, clear.
- Good chemical and physical properties
- For applications that do not involve high thermal loads
Ideal for sampling, storing and transporting powders and pastes.