Cold storage overalls, IGLOO II
Supplier: DELTA PLUS
Certificates
About this item
These overalls made of 85% polyester and 15% cotton feature a nylon lining and a Thinsulate™ padding. Ideal for extreme cold environments.
- Fixed hood with polar fleece flining and Thinsulate™ padding
- Front zip closure with flap
- Two chest pockets with velcro®, pen pocket, phone pocket
- Two bottom pockets with flap and velcro®
- Elasticated waist at the back, elasticated cuffs
- Bottom legs with inside zip and flap, adjustable by velcro®
Certifications: ENV 342 (2-3)
Specifications
- Catalog No:
- DELPIGLO2BLXG
- DELPIGLO2BLXX
- Modello:
- IGLOO II
- IGLOO II
- Applicazioni per abiti:
- Industrial
- Industrial
- Risk area:
- Cold
- Cold
- Genere:
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Tessuto:
- 85% polyester, 15% cotton
- 85% polyester, 15% cotton