Softshell jackets, Oregon, TK40
Nome del fornitore: Portwest
Informazioni su questo articolo
These 2-layer softshell jackets are made of 100% polyester bonded with polyester micro polar fleece. The jackets are wind- and showerproof.
- Two side pockets, zipped mobile phone pocket (chest)
- Half-elasticated cuffs with Velcro adjustment
Colour: Black
Weight: 250 g/m²
6 varianti
Product Details & Documents
These 2-layer softshell jackets are made of 100% polyester bonded with polyester micro polar fleece. The jackets are wind- and showerproof.
- Two side pockets, zipped mobile phone pocket (chest)
- Half-elasticated cuffs with Velcro adjustment
Colour: Black
Weight: 250 g/m²
Specifications for Product Family
Specifiche
- Catalog No:
- PRTWTK40BKRS
- PRTWTK40BKRM
- PRTWTK40BKRL
- PRTWTK40BKRXL
- PRTWTK40BKRXXL
- PRTWTK40BKRXXXL
- Modello:
- Oregon TK40
- Oregon TK40
- Oregon TK40
- Oregon TK40
- Oregon TK40
- Oregon TK40
- Applicazioni per abiti:
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Risk area:
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Genere:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Tipo di chiusura:
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Tessuto:
- 100% polyester bonded with polyester micro polar fleece
- 100% polyester bonded with polyester micro polar fleece
- 100% polyester bonded with polyester micro polar fleece
- 100% polyester bonded with polyester micro polar fleece
- 100% polyester bonded with polyester micro polar fleece
- 100% polyester bonded with polyester micro polar fleece
- Peso:
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
Specifiche
Product Family Options
Informazioni sul prodotto
- ColoreTaglia dell'abbigliamentoVestibilità del capoDisponibilitàPrice
- Specifiche:
More
Specifications
Cat. No.PRTWTK40BKRSModelloOregon TK40Applicazioni per abitiOutdoorRisk areaColdGenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessuto100% polyester bonded with polyester micro polar fleecePeso250 g/m² - Specifiche:
More
Specifications
Cat. No.PRTWTK40BKRMModelloOregon TK40Applicazioni per abitiOutdoorRisk areaColdGenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessuto100% polyester bonded with polyester micro polar fleecePeso250 g/m² - Specifiche:
More
Specifications
Cat. No.PRTWTK40BKRLModelloOregon TK40Applicazioni per abitiOutdoorRisk areaColdGenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessuto100% polyester bonded with polyester micro polar fleecePeso250 g/m² - Specifiche:
More
Specifications
Cat. No.PRTWTK40BKRXLModelloOregon TK40Applicazioni per abitiOutdoorRisk areaColdGenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessuto100% polyester bonded with polyester micro polar fleecePeso250 g/m² - Specifiche:
More
Specifications
Cat. No.PRTWTK40BKRXXLModelloOregon TK40Applicazioni per abitiOutdoorRisk areaColdGenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessuto100% polyester bonded with polyester micro polar fleecePeso250 g/m² - Specifiche:
More
Specifications
Cat. No.PRTWTK40BKRXXXLModelloOregon TK40Applicazioni per abitiOutdoorRisk areaColdGenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessuto100% polyester bonded with polyester micro polar fleecePeso250 g/m²