ContactUsLinkComponent
Wireless monitoring systems, ready-to-use kits, Sensor2Web™
Wireless monitoring systems, ready-to-use kits, Sensor2Web™
Catalog # 620-1992
Supplier:  Rmoni
Wireless monitoring systems, ready-to-use kits, Sensor2Web™
Catalog # 620-1992
Supplier:  Rmoni
Specifications
  • Descrizione:
    Monitoring wireless system of temperature in meat processors, industrial bakeries and fresh vegetables, FRESH-MONI Sensor2Web kit
  • Conf:
    1 KIT
Specifications
About this item
Sensor2Web is a wireless monitoring system for continuous recording and monitoring of critical parameters.
  • Management of configurations in each department
  • Access to the data via a standard web browser
  • Data is stored in the cloud (sensor2web) or on the server (sensor4lab)
  • Reports for controlling organisations, auditors or customers are ready to use and self-configurable
  • Receive alarms and respond immediately; alarms via SMS, email or voice mail
Sensor2Web makes it possible to follow close and extended, with rapid reaction when changes occur. With this unique combination of wireless technology and web-based software, it also provides a cost-effective and time-saving alternative to manual measurements. Some configured kits are available for a quick start and rapid startup.
Typical applications include the monitoring of temperatures in laboratory equipment such as refrigerators, freezers, incubators; temperature of pharmaceutical products in the Cold Supply Chain according to GDP, ambient conditions in clean rooms, and the temperature of food during production and transport in central kitchens and hospitals.
Delivery: These kits include a gateway, endpoints and sensors (according to their application), and the fee for 1 year.