Spill response kit for the laboratory, PIG® HAZ-MAT
Référence 121-0568
Fournisseur: NEW PIG
Spécifications
- Descrizione:Spill response kit for the laboratory
- Conf:1 KIT
Spécifications
À propos de cet article
Small spill kit suitable chemical spills.
- Compatible with the most acids and bases including 98% sulphuric acid and 30% sodium hydroxide
- Sock and mats for containment and absorption
- Pair of disposable gloves and bags for rapid response and disposal
Livraison: Supplied with 9× absorbent mats (380×510 mm) ; 1× absorbent sock (1,2 m) ; 1× pair 'Silver Shield' chemical gloves ; and 2× plastic disposal bags,1× warning sign and disposal guide.