Barcode reader, Nunc™
Catalog # NUNC330050
Supplier: Thermo Fisher Scientific
Specifications
- Descrizione:Barcode scanner, for use with any USB port
- Interfaccia:USB
- Conf:1
Specifications
About this item
This durable barcode reader is ideal for simple track sampling. It is for use with any USB slot.
- 'Plug and Play' function can automatically switch to read codes: 128A, 128B, 128C, 39 and interleave ²/₅ Convenient and easy to use with an Excel spread sheet
- Scanner is a contact scanner which means it must be in contact with the barcode to read