Orders LinkContactUsLinkComponent
Container, rectangular, with handle | Avantor
Container, rectangular, with handle
10 l, 187×288×273 mm, -
Katalognr. 216-1604
Anbieter:   HK PLASTICS
Container, rectangular, with handle
Katalognr. 216-1604
Anbieter:  HK PLASTICS
Container, rectangular, with handle
Container, rectangular, with handle
10 l, 187×288×273 mm, -
Preis
4 250.00 SFr.
Einloggen für Verfügbarkeit
1 * 600 ST
Product Details & Documents
PP/PE.
  • Increase storage capacity, reduce the transport volume and the required shelf space
  • All materials are food grade approved and coloured master batch, cadmium free
  • Designed to be nestable and interlocking
Robust rectangular containers, which are easy to open and (re)close. Strong, ergonomic handle grips making them easy to transport.
Produced under BRC/IoP regulation, according to HACCP standards.
Technische Daten
  • Tipo:
    -
  • Capacità:
    10 l
  • L×P×A:
    187×288×273 mm