Handheld refractometers
0 - 80 °Brix, 0,5 °Brix
Codice catalogo 635-0081
Nome del fornitore: EUROMEX
Vai a
Product Details & Documents
Single dual or triple scales, analogue, handheld refractometers.
- Automated temperature compensation (ATC)
- Calibrated at 20 °C except for RF.5190
- Requires only a few drops of liquid
Specifiche
- Range di misura:0 - 80 °Brix
- Seq:07
- Precisione:0,5 °Brix