Cleanroom sleeve protectors, Kimtech™ A5
Catalog # 113-7733
Supplier: KIMBERLY CLARK
Specifications
- Sterili:+
- Applicazioni per abiti:Cleanroom
- Colore:bianco
- Lunghezza:457 mm
- Environmentally Preferable:Riduzione degli sprechi
- Conf:100 PAIR
- Classe di cleanroom:100/ISO 5
Specifications
About this item
These sterile sleeve protectors are made of durable SMS fabric which provides a cloth-like feel. A highly visible blue line along the inside of the sleeve signals the proper place to grasp while donning, helping workers avoid touching the outside.
- Thumb loop keeps sleeve from rolling back and exposing arm or wrist
- Tunnelled elastic and bound seams
- Triple-bagged
Suitable for Class 100, ISO Class 5 or higher cleanrooms.
Certifications: PPE Cat I (EU) 2016/425.