Orders LinkContactUsLinkComponent
Bosshead, cross type
Bosshead, cross type
Bosshead, cross type
Bosshead made of tempered cast iron, powder-coated.
  • For rods up to 16 mm Ø
  • With brass thumb or stainless steel safety heart screws
  • Thread: M8
Weight: 180 g
Product Family Options
  • Descrizione
    Larghezza di serraggio
    Materiale
  • With brass thumb screws
    0 - 16 mm
    Ghisa temprata
    1 * 1 ST
    1 * 1 ST - 45.10 SFr.
    45.10 SFr.
    1 * 1 ST
       45.10 SFr.
  • With safety heart screws
    0 - 16 mm
    Ghisa temprata
    1 * 1 ST
    1 * 1 ST - 91.10 SFr.
    91.10 SFr.
    1 * 1 ST
       91.10 SFr.