Fume hood, benchtop, small, SP Bel-Art
Catalog # BELAH500000002
Supplier: Bel-Art Products, a Part of SP
Specifications
- Esterno L×P×A:690×330×560 mm
- Descrizione:Benchtop, front opening 560×300 mm
- Conf:1
Specifications
About this item
Benchtop fume hood designed for small facilities or installations in which a number of fume hoods are needed, this unit is mainly used in schools, junior colleges and universities.
- Easy to clean
- Bottom gap allows for continuous air flow
- One-piece shell moulded of chemical resistant polyethylene with rounded inside corners
- Clear acrylic sash slides on a track featuring stop pins for open, closed or intermediate positions