Ball valves
Ball valves, with an inner and outer thread, PP, 3/4" NW 20
Codice catalogo 229-7490
Nome del fornitore: SEMADENI
Vai a
Product Details & Documents
Housing and ball made of PP, ball seal made of PTFE. These ball valves has a easy running, strong industrial design.
- Operating pressure of 6 kg/cm² at max. 80 °C
- Excellent chemical resistance
Covered both sides with gas thread for direct installation or via connection for hose.
Specifiche
- Descrizione:Ball valves, with an inner and outer thread, PP, 3/4" NW 20