HPLC columns, Discovery RP-AmideC16, Supelco®
Supplier: Merck
Certificates
About this item
These columns with excellent retention and resolution of polar compounds are less hydrophobic than C18 columns.
- Unique selectivity compared to C18
- Excellent peak shape and efficiency
- Compatible with 100% aqueous mobile phases
Specifications
- Catalog No:
- 505013-40
- 505064-30
- Carico di carbonio:
- 11%
- 11%
- Con endcapping:
- Sì
- Sì
- Range di pH:
- 2 - 8
- 2 - 8
- Area superficiale:
- 200 m²/g
- 200 m²/g
- Codice USP:
- L60
- L60