Microcentrifuge tubes
Codice catalogo NEOL7-4590
Nome del fornitore: NEOLAB
Specifiche
- Confezione:1000 pcs/pk
- Tipo:with thread, without screw cap
- Versione:non graduated, skirted, non sterile
- Capacità:0,5 ml
- Colore:ambra
- Conf:1.000
Specifiche
Informazioni su questo articolo
PP, amber coloured.
- Ungraduated
- Amber coloured to protect light sensitive substances
- Excellent chemical resistance
- Excellent temperature resistance
With thread.
Ordering information: Screw caps must be ordered seperately.