Lab markers, water resistant
Codice catalogo NEOL2-5040
Nome del fornitore: NEOLAB
Specifiche
- Colore:nero
- Descrizione:Laboratory marker, water resistant, fine tip
- Conf:1
Specifiche
Informazioni su questo articolo
Markers write on all materials used in a laboratory (as glass, plastics, porcelain, metal, wood, paper, etc.) and perform equally well on dry or moist, cold or warm surfaces.
- Fine tip
- Ink won't bleed when subjected to water or alcohol during normal use
- Yet can be washed off