Orders LinkContactUsLinkComponent
Safety caps
Safety cap II (includes 1 PP blue basic cap with 2 ports and 2 shut-off valves, GL 45, with PTFE cone, 2 PFA fittings, 3,2 mm (1 blue, 1 red), 2 white PTFE fittings, 3,2 mm, 1 venting valve, including air filter, 1 μm, PTFE membrane), for GL 45 threaded bottles
  HEWL5043-0224
Safety caps
  HEWL5043-0224
Safety caps
Safety caps
Safety cap II (includes 1 PP blue basic cap with 2 ports and 2 shut-off valves, GL 45, with PTFE cone, 2 PFA fittings, 3,2 mm (1 blue, 1 red), 2 white PTFE fittings, 3,2 mm, 1 venting valve, including air filter, 1 μm, PTFE membrane), for GL 45 threaded bottles
1 * 1 ST
These safety caps have been designed for optimal sealing with an integrated exhaust valve providing pressurisation during solvent extraction and allowing proper solvent flow to your HPLC system.
  • Caps rotate freely, preventing tube twisting during bottle exchange
  • Constructed of PTFE and PFA for high chemical resistance
The exhaust valve contains a PTFE membrane to prevent contamination of your solvents from dirt and dust particles.
  • Descrizione:
    Safety cap II (includes 1 PP blue basic cap with 2 ports and 2 shut-off valves, GL 45, with PTFE cone, 2 PFA fittings, 3,2 mm (1 blue, 1 red), 2 white PTFE fittings, 3,2 mm, 1 venting valve, including air filter, 1 μm, PTFE membrane), for GL 45 threaded bottles
  • Seq:
    01
Accessories
  •  
    Descrizione
  • HEWL5043-0229
    Fittings, PFA, for 1,6 mm tube
    1 * 5 ST
  •  
    Descrizione
  • HEWL5043-0238
    Collector, 3 ports
    1 * 1 ST
  •  
    Descrizione
  • HEWL5043-0239
    Adapter, for 2×3,2 mm tubes
    1 * 1 ST
  •  
    Descrizione
  • HEWL5043-0255
    Fittings, PFA, for 3,2 mm tube
    1 * 6 ST