Electron multipliers
Anbieter: AGILENT HEWLETT PACKARD
Zertifikate
Über diesen Artikel
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Anbieter: VWR Chemicals
Acetone ≥99%, TECHNICAL
Anbieter: VWR Chemicals
VWR®, Snap Cap Vials
Anbieter: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Anbieter: VWR Chemicals
VWR®, Short Thread Vials ND9
Anbieter: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Anbieter: VWR Collection
Acqua, HiPerSolv CHROMANORM® per HPLC
Anbieter: VWR Chemicals
Metanolo ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grado gradiente per HPLC
Anbieter: VWR Chemicals
VWR®, Screw Neck Vials ND13
Anbieter: VWR Collection
Etanolo assoluto ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Anbieter: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Anbieter: VWR Collection
Cleanroom Waste Receptacles
Anbieter: Palbam