Orders LinkContactUsLinkComponent
Iris scissors, Aesculap®
 :   Aesculap
Iris scissors, Aesculap®
 :  Aesculap
Iris scissors, Aesculap®
Product Family Options
  • Tipo
    Lunghezza
  • Strabismus, angled, sharp/with knob
    115 mm
    1 * 1 ST
    1 * 1 ST - SFr. 149.00
    SFr. 149.00
    1 * 1 ST
       SFr. 149.00
  • Straight, blunt/blunt (pointed probe)
    145 mm
    1 * 1 ST
    1 * 1 ST - SFr. 144.00
    SFr. 144.00
    1 * 1 ST
       SFr. 144.00
  • Straight, sharp/sharp
    95 mm
    1 * 1 ST
    1 * 1 ST - SFr. 64.30
    SFr. 64.30
    1 * 1 ST
       SFr. 64.30
  • Curved, sharp/sharp
    95 mm
    1 * 1 ST
    1 * 1 ST - SFr. 69.90
    SFr. 69.90
    1 * 1 ST
       SFr. 69.90
  • Straight, sharp/sharp
    110 mm
    1 * 1 ST
    1 * 1 ST - SFr. 58.30
    SFr. 58.30
    1 * 1 ST
       SFr. 58.30
  • Curved, sharp/sharp
    110 mm
    1 * 1 ST
    1 * 1 ST - SFr. 63.80
    SFr. 63.80
    1 * 1 ST
       SFr. 63.80
  • Straight, sharp/sharp
    120 mm
    1 * 1 ST
    1 * 1 ST - SFr. 68.50
    SFr. 68.50
    1 * 1 ST
       SFr. 68.50
  • Curved, sharp/sharp
    120 mm
    1 * 1 ST
    1 * 1 ST - SFr. 75.70
    SFr. 75.70
    1 * 1 ST
       SFr. 75.70
  • Straight, sharp/sharp
    90 mm
    1 * 1 ST
    1 * 1 ST - SFr. 48.60
    SFr. 48.60
    1 * 1 ST
       SFr. 48.60
  • Curved, sharp/sharp
    90 mm
    1 * 1 ST
    1 * 1 ST - SFr. 55.00
    SFr. 55.00
    1 * 1 ST
       SFr. 55.00
  • Straight, sharp/sharp
    115 mm
    1 * 1 ST
    1 * 1 ST - SFr. 55.20
    SFr. 55.20
    1 * 1 ST
       SFr. 55.20
  • Curved, sharp/sharp
    115 mm
    1 * 1 ST
    1 * 1 ST - SFr. 58.30
    SFr. 58.30
    1 * 1 ST
       SFr. 58.30
  • Straight, sharp/sharp
    110 mm
    1 * 1 ST
    1 * 1 ST - SFr. 53.50
    SFr. 53.50
    1 * 1 ST
       SFr. 53.50
  • Curved, sharp/sharp
    110 mm
    1 * 1 ST
    1 * 1 ST - SFr. 56.20
    SFr. 56.20
    1 * 1 ST
       SFr. 56.20
  • Straight, sharp/blunt
    110 mm
    1 * 1 ST
    1 * 1 ST - SFr. 66.40
    SFr. 66.40
    1 * 1 ST
       SFr. 66.40
  • Curved, sharp/blunt
    110 mm
    1 * 1 ST
    1 * 1 ST - SFr. 66.40
    SFr. 66.40
    1 * 1 ST
       SFr. 66.40
  • Angled, sharp/sharp
    110 mm
    1 * 1 ST
    1 * 1 ST - SFr. 121.00
    SFr. 121.00
    1 * 1 ST
       SFr. 121.00
  • Suture scissors, straight, sharp/sharp
    120 mm
    1 * 1 ST
    1 * 1 ST - SFr. 65.50
    SFr. 65.50
    1 * 1 ST
       SFr. 65.50
  • Suture scissors, curved, sharp/sharp
    120 mm
    1 * 1 ST
    1 * 1 ST - SFr. 72.50
    SFr. 72.50
    1 * 1 ST
       SFr. 72.50
  • Suture scissors, curved, blunt/blunt
    120 mm
    1 * 1 ST
    1 * 1 ST - SFr. 72.50
    SFr. 72.50
    1 * 1 ST
       SFr. 72.50
  • Suture scissors, straight, sharp/sharp
    130 mm
    1 * 1 ST
    1 * 1 ST - SFr. 71.50
    SFr. 71.50
    1 * 1 ST
       SFr. 71.50
  • Suture scissors, curved, sharp/sharp
    130 mm
    1 * 1 ST
    1 * 1 ST - SFr. 78.00
    SFr. 78.00
    1 * 1 ST
       SFr. 78.00
  • Suture scissors, straight, blunt/blunt
    130 mm
    1 * 1 ST
    1 * 1 ST - SFr. 86.00
    SFr. 86.00
    1 * 1 ST
       SFr. 86.00
  • Suture scissors, curved, blunt/blunt
    130 mm
    1 * 1 ST
    1 * 1 ST - SFr. 95.30
    SFr. 95.30
    1 * 1 ST
       SFr. 95.30
  • Chadwick scissors, strongly curved, sharp/sharp
    115 mm
    1 * 1 ST
    1 * 1 ST - SFr. 142.00
    SFr. 142.00
    1 * 1 ST
       SFr. 142.00
  • La Grange, curved, sharp/sharp
    110 mm
    1 * 1 ST
    1 * 1 ST
  • Joseph scissors, straight, sharp/sharp
    150 mm
    1 * 1 ST
    1 * 1 ST - SFr. 91.80
    SFr. 91.80
    1 * 1 ST
       SFr. 91.80
  • Kelly scissors, straight, sharp/sharp
    175 mm
    1 * 1 ST
    1 * 1 ST - SFr. 114.00
    SFr. 114.00
    1 * 1 ST
       SFr. 114.00
  • Kelly scissors, curved, sharp/sharp
    175 mm
    1 * 1 ST
    1 * 1 ST - SFr. 121.00
    SFr. 121.00
    1 * 1 ST
       SFr. 121.00
  • Gum scissors, curved, sharp/sharp
    120 mm
    1 * 1 ST
    1 * 1 ST
  • Gum scissors, curved, sharp/sharp
    120 mm
    1 * 1 ST
    1 * 1 ST
  • Quinby scissors, curved, sharp/sharp
    130 mm
    1 * 1 ST
    1 * 1 ST
  • Locklin scissors, angled to side, sharp/sharp (1 blade toothed)
    155 mm
    1 * 1 ST
    1 * 1 ST
  • Gum scissors, Goldman-Fox, curved, sharp/sharp (1 blade toothed)
    130 mm
    1 * 1 ST
    1 * 1 ST
  • Gum scissors, Goldman-Fox, angled on flat, sharp/sharp (1 blade toothed)
    125 mm
    1 * 1 ST
    1 * 1 ST
  • Perwitzschky Salival, curved, sharp/with knob
    100 mm
    1 * 1 ST
    1 * 1 ST
  • Tonsil scissors, Dean, angled on flat, blunt/blunt
    170 mm
    1 * 1 ST
    1 * 1 ST
  • Straight, sharp/sharp
    110 mm
    1 * 1 ST
    1 * 1 ST - SFr. 73.80
    SFr. 73.80
    1 * 1 ST
       SFr. 73.80