Orders LinkContactUsLinkComponent
Torion glass joints, SVL®
Torion glass joints, SVL®
Nome del fornitore:   SciLabware
Torion glass joints, SVL®
Nome del fornitore:  SciLabware
Certificates
Torion glass joints, SVL®
Torion glass joints, SVL®
3 varianti
Product Details & Documents
Jointing system for glassblowers to make connections between tubing and glassware.
  • Excellent chemical resistance
  • Excellent temperature resistance
Glass body has an internal screwthread into which is fitted the rubber sealing ring sheathed with PTFE. Tubing is pushed through the screwthread fitting, which is then threaded onto the glass body. As the fitting is tightened, the sealing ring is compressed to form a seal between the tubing and the body. Only glass and PTFE come in to contact and provide a chemical resistant pathway.
Informazioni per l'ordine: Complete joint comprises borosilicate glass body, glass filled polypropylene fitting and rubber/PTFE sealing ring.
Product Family Options
Informazioni sul prodotto
  • Disponibilità
    Price
  • BIBB721-01
    Complete jointing system for tubing 4,8 - 5,2 mm O.Ø, O.Ø×L 14×8 mm shank, O.Ø×H 21×27 mm body
    Complete jointing system for tubing 4,8 - 5,2 mm O.Ø, O.Ø×L 14×8 mm shank, O.Ø×H 21×27 mm body
    Log in per disponibilità
    Log in per disponibilità
    1 * 10 ST
    1 * 10 ST - 506.00 SFr.
    506.00 SFr.
    1 * 10 ST
       506.00 SFr.
  • BIBB721-02
    Complete jointing system for tubing 7,6 - 8,4 mm O.Ø, O.Ø×L 17×9 mm shank, O.Ø×H 26×32 mm body
    Complete jointing system for tubing 7,6 - 8,4 mm O.Ø, O.Ø×L 17×9 mm shank, O.Ø×H 26×32 mm body
    Log in per disponibilità
    Log in per disponibilità
    1 * 10 ST
    1 * 10 ST - 641.00 SFr.
    641.00 SFr.
    1 * 10 ST
       641.00 SFr.
  • BIBB721-03
    Complete jointing system for tubing 13,6 - 14,11 mm O.Ø, O.Ø×L 25×15 mm shank, O.Ø×H 37×42 mm body
    Complete jointing system for tubing 13,6 - 14,11 mm O.Ø, O.Ø×L 25×15 mm shank, O.Ø×H 37×42 mm body
    Log in per disponibilità
    Log in per disponibilità
    1 * 10 ST
    1 * 10 ST - 841.00 SFr.
    841.00 SFr.
    1 * 10 ST
       841.00 SFr.
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Sealing rings, for tubing 13,6 - 14,4 mm
    Sealing rings, for tubing 13,6 - 14,4 mm
    BIBB701-24
    Torion joints
    Log in per disponibilità
    1 * 10 ST - SFr. 187.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Fittings, PP, bore Ø 14,7 mm
    Fittings, PP, bore Ø 14,7 mm
    BIBB720-03
    Torion glass bodies and sealing rings (rubber/PTFE)
    Log in per disponibilità
    1 * 10 ST - SFr. 176.00
Informazioni sul prodotto
  • Codice catalogo
    Per
    Disponibilità
    Price
  • Sealing rings, for tubing 7,6 - 8,4 mm
    Sealing rings, for tubing 7,6 - 8,4 mm
    BIBB701-23
    Torion joints
    Log in per disponibilità
    1 * 10 ST - SFr. 114.00