Folded gown dispensers
Supplier: Palbam
Certificates
About this item
These dispensers are suitable for packs of folded gowns in cleanrooms, for use in industries such as pharmaceutical, biotechnology, medical device, microelectronics and nanotechnology.
- High quality design
- Can be wall-mounted
- Hold a pack of folded gown
- Stainless steel 304 electropolished finish
Certifications: ISO certified
Delivery: Typically available from stock for direct shipment from the supplier.
Specifications
- Catalog No:
- 113-8177
- 113-8173
- PPE type:
- Indumenti usa e getta
- Indumenti usa e getta
- Materiale:
- Acciaio inossidabile
- Acciaio inossidabile
- Tipo di dispenser:
- Montabile a parete
- Montabile a parete
- L×P×A:
- 410×310×750 mm
- 350×250×750 mm