Orders LinkContactUsLinkComponent
Safety caps, GL 45 threads
Nome del fornitore:   VICI JOUR RESEARCH
Safety caps, GL 45 threads
Nome del fornitore:  VICI JOUR RESEARCH
Certificates
Safety caps, GL 45 threads
9 varianti
Product Details & Documents
GL 45 safety caps seal to the bottle with an O-ring (EPDM) which is resistant to commonly used HPLC solvents. If improved chemical resistance is required (e.g. for halogenated hydrocarbons), order an FEP-coated O-ring separately.
  • Built-in stopcocks for easy interruption of solvent lines
  • Fit all bottles or canisters with DIN50 thread
  • Prevent evaporation of volatile compounds
  • Stopcocks rated up to 72 psi gas at 25 °C
 : A bottom-sealing barbed adapter to connect ¹/₈" (3,2 mm) I.Ø tubing can be ordered separately.
Product Family Options
Informazioni sul prodotto
  • Filettatura
    Connettore
    Materiale
    Disponibilità
    Price
  • JOURJR-S-11011
    Safety cap, with stopcock, EPDM O-ring, nuts, ferrules and colour-sleeve adapters
    Safety cap, with stopcock, EPDM O-ring, nuts, ferrules and colour-sleeve adapters
    GL 45
    Two ports with 1/4"-28 thread for 1/8" or 1/16" O.Ø tubing
    PP
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 315.00
    SFr. 315.00
    1 * 1 ST
       SFr. 315.00
  • JOURJR-S-11012
    Safety cap, with stopcock, EPDM O-ring, nuts, ferrules and colour-sleeve adapters
    Safety cap, with stopcock, EPDM O-ring, nuts, ferrules and colour-sleeve adapters
    GL 45
    Three ports with 1/4"-28 thread for 1/8" or 1/16" O.Ø tubing
    PP
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 394.00
    SFr. 394.00
    1 * 1 ST
       SFr. 394.00
  • JOURJR-S-11013
    Safety cap, with stopcock, EPDM O-ring, nuts, ferrules and colour-sleeve adapters
    Safety cap, with stopcock, EPDM O-ring, nuts, ferrules and colour-sleeve adapters
    GL 45
    Four ports with 1/4"-28 thread for 1/8" or 1/16" O.Ø tubing
    PP
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 465.00
    SFr. 465.00
    1 * 1 ST
       SFr. 465.00
Safety cap kits
Informazioni sul prodotto
  • Filettatura
    Connettore
    Materiale
    Disponibilità
    Price
  • JOURJR-S-11111
    Safety cap kit, includes 1× safety cap, with fittings, 1× 1 l safety coated glass bottle, 3 m PFA tubing, 1× 5 µm mobile phase filter, nuts and ferrules
    Safety cap kit, includes 1× safety cap, with fittings, 1× 1 l safety coated glass bottle, 3 m PFA tubing, 1× 5 µm mobile phase filter, nuts and ferrules
    GL 45
    Two ports with 1/4"-28 thread for 1/8" or 1/16" O.Ø tubing
    PP
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 465.00
    SFr. 465.00
    1 * 1 ST
       SFr. 465.00
  • JOURJR-S-11112
    Safety cap kit, includes 1× safety cap, with fittings, 1× 1 l safety coated glass bottle, 3 m PFA tubing, 1× 5 µm mobile phase filter, nuts and ferrules
    Safety cap kit, includes 1× safety cap, with fittings, 1× 1 l safety coated glass bottle, 3 m PFA tubing, 1× 5 µm mobile phase filter, nuts and ferrules
    GL 45
    Three ports with 1/4"-28 thread for 1/8" or 1/16" O.Ø tubing
    PP
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 601.00
    SFr. 601.00
    1 * 1 ST
       SFr. 601.00
  • JOURJR-S-11113
    Safety cap kit, includes 1× safety cap, with fittings, 1× 1 l safety coated glass bottle, 3 m PFA tubing, 1× 5 µm mobile phase filter, nuts and ferrules
    Safety cap kit, includes 1× safety cap, with fittings, 1× 1 l safety coated glass bottle, 3 m PFA tubing, 1× 5 µm mobile phase filter, nuts and ferrules
    GL 45
    Four ports with 1/4"-28 thread for 1/8" or 1/16" O.Ø tubing
    PP
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 734.00
    SFr. 734.00
    1 * 1 ST
       SFr. 734.00
  • JOURJR-S-11211
    Safety cap kit, includes 1× safety cap, with fittings, 1× 2 l safety coated glass bottle, 3 m PFA tubing, 1× 5 µm mobile phase filter, nuts and ferrules
    Safety cap kit, includes 1× safety cap, with fittings, 1× 2 l safety coated glass bottle, 3 m PFA tubing, 1× 5 µm mobile phase filter, nuts and ferrules
    GL 45
    Two ports with 1/4"-28 thread for 1/8" or 1/16" O.Ø tubing
    PP
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 546.00
    SFr. 546.00
    1 * 1 ST
       SFr. 546.00
  • JOURJR-S-11212
    Safety cap kit, includes 1× safety cap, with fittings, 1× 2 l safety coated glass bottle, 3 m PFA tubing, 1× 5 µm mobile phase filter, nuts and ferrules
    Safety cap kit, includes 1× safety cap, with fittings, 1× 2 l safety coated glass bottle, 3 m PFA tubing, 1× 5 µm mobile phase filter, nuts and ferrules
    GL 45
    Three ports with 1/4"-28 thread for 1/8" or 1/16" O.Ø tubing
    PP
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 683.00
    SFr. 683.00
    1 * 1 ST
       SFr. 683.00
  • JOURJR-S-11213
    Safety cap kit, includes 1× safety cap, with fittings, 1× 2 l safety coated glass bottle, 3 m PFA tubing, 1× 5 µm mobile phase filter, nuts and ferrules
    Safety cap kit, includes 1× safety cap, with fittings, 1× 2 l safety coated glass bottle, 3 m PFA tubing, 1× 5 µm mobile phase filter, nuts and ferrules
    GL 45
    Four ports with 1/4"-28 thread for 1/8" or 1/16" O.Ø tubing
    PP
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 820.00
    SFr. 820.00
    1 * 1 ST
       SFr. 820.00
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • O-ring, FEP coated, GL45 thread
    O-ring, FEP coated, GL45 thread
    JOURJR-S-40008
    Log in per disponibilità
    1 * 1 ST - SFr. 49.10