Anion exchange HPLC columns, Dionex™, IonPac™ AS9-SC
Anbieter: DIONEX
Zertifikate
Über diesen Artikel
AS9-SC columns are used for the separations of oxyhalides and inorganic anions including fluoride, chlorite, bromate, chloride, nitrite, bromide, chlorate, nitrate, phosphate, and sulphate in drinking water, wastewater, groundwater, and other sample matrices.
- Fast analysis in similar concentrations
- High-capacity for trace bromate in ozonated drinking water
Technische Daten
- Catalog No:
- DION043186
- DION043185
- Pressione max.:
- 4000 psi
- 4000 psi
- Range di pH:
- 2 - 11
- 2 - 11
Technische Daten
Frequently Bought Together