Cut resistant gloves, Sharpflex Liner
9, Giallo/verde, 250 mm
Catalog # DALL2232522/9
Supplier: HONEYWELL SAFETY
Jump to:
Product Details & Documents
These gloves are knitted from composite and Kevlar® yarn.
- Knitting technique provides the glove with excellent fit and high dexterity
- Kevlar® yarn protects against contact heat and cuts
- Plated polyamide inside: Limit the contact of the skin with composite fibre
- Ambidextrous
Ideal for handling heavy parts with high cut risks in dry environments.
Certifications: EN 388:2016 (2X4XC), EN 407 (X1XXXX); Cat. II
Specifications
- Dimensione del guanto:9
- Colore:Giallo/verde
- Lunghezza:250 mm
- Seq:03
- Monouso/riutilizzabile:riutilizzabile
- Norma EN:EN 388:2016 (2X4XC), EN 407 (X1XXXX)
- Cat. No.:DALL2232522/9
- Ambidestro/mano specifico:ambidestro
- Modello:Liner
- Tipo polsino:Maglia elastica
- Calibro:13