ContactUsLinkComponent
Disposal box for blades, SafeBox
Catalog # MARO108000.00
Supplier:  MARTOR ARGENTAX
undefined
Disposal box for blades, SafeBox
Catalog # MARO108000.00
Supplier:  MARTOR ARGENTAX
Specifications
  • Descrizione:
    SafeBox, for the deposition of used blades up to a length of 62 mm
  • Conf:
    1
Specifications
About this item
This box is designed for the safe deposition of 'used' snap-off blade segments, blade ends, straight/hook/industrial cutter/deburring scalpel and hobby crafts blades.
Absolutely safe and inaccessible to fingers.