Dispenser for hand towel rolls, AQUARIUS* SLIMROLL*
Codice catalogo KIMB6953
Nome del fornitore: KIMBERLY CLARK
Specifiche
- Descrizione:AQUARIUS* SLIMROLL* dipsenser for hand towel rolls, with window, white
- L×P×A:326×201×360 mm
- Conf:1
- Per:Towel rolls
Specifiche
Informazioni su questo articolo
This dispenser enables single sheet dispensing of KLEENEX® and SCOTT* rolled hand towels, without the need to touch the dispenser. Ideal for use in high traffic environments such as offices, food processing environments and public venues (airports and sports stadiums).
- Helps to reduce contamination and the spread of germs
- With easy maintenance and minimal servicing requirements
- With push button or key lock
- Curved design with an easy-clean finish and no dirt or dust traps
- Stub-roll facility holds rolls up to 165-m length, this provides up to 600 sheets (25 cm width)