Cooling racks
Cooling rack, acrylic, for 0,2 ml PCR vials, 130×90×30 mm
Katalognr. KOEHA-1070
Anbieter: KOEHLER TECHNISCHE PRODUKTEN
Direkt zu
Product Details & Documents
PCR vials may be kept at a temperature of 0 °C for at least 30 min if the cooler is cooled down to –20 °C for approximately 12 h.
Technische Daten
- Descrizione:Cooling rack, acrylic, for 0,2 ml PCR vials
- L×P×A:130×90×30 mm