Orders LinkContactUsLinkComponent
Lockout device for plug connections
Lockout device for plug connections
Nome del fornitore:   Brady
Lockout device for plug connections
Nome del fornitore:  Brady
Certificates
Lockout device for plug connections
Lockout device for plug connections
2 varianti
Product Details & Documents
This lockout device is highly effective against the unauthorised or incorrect use of electrical equipment.
  • Made from PP: Temperature resistant from −20 to +80 °C
  • Small device accommodates two padlocks, large device four padlocks
  • Easy to use
  • Secures all types of plugs
Min. shackle clearance: 20,00 mm
Max. shackle diameter: 7,00 mm
Cable diameter: 10 mm (small) or 22 mm (large)
Product Family Options
Informazioni sul prodotto
  • Tipo
    Colore
    L×P×A
    Disponibilità
    Price
  • Lockout device, large, for plug connections with max. Ø 22 mm
    rosso
    178×84×117 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 30.40
    SFr. 30.40
    1 * 1 ST
       SFr. 30.40
  • Lockout device, small, for plug connections with max. Ø 10 mm
    rosso
    88,9×43,2×80 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 22.10
    SFr. 22.10
    1 * 1 ST
       SFr. 22.10