ContactUsLinkComponent
Test tube holder, stainless steel
Test tube holder, stainless steel
Katalognr. 241-0233
Anbieter:  HAMMACHER KARL
undefined
Test tube holder, stainless steel
Katalognr. 241-0233
Anbieter:  HAMMACHER KARL
Technische Daten
  • Lunghezza:
    135 mm
  • Conf:
    1
Technische Daten
Über diesen Artikel
Test tube holder made from WIRONIT steel (18/12 stainless steel with 18% chrome and 10 to 12% nickel).