Disarticulated half human skeleton, loosely articulated hand and foot
Codice catalogo BINH1000024
Nome del fornitore: 3B Scientific
Specifiche
- Descrizione:Disarticulated half human skeleton, loosely articulated hand and foot
- Conf:1
Specifiche
Informazioni su questo articolo
This half skeleton has the hand and foot loosely articulated on nylon cord for study of more realistic movement. This replica of a disarticulated half human skeleton is complete with mounted skull, sternum, hyoid, and spinal column. This disarticulated skeleton is great for study of the anatomy of the human skeleton and any scientific anatomy studies.
- Comes in a sturdy partitioned cardboard storage box
- High quality model with realistic anatomic detail
Dimensions: 48,5×27×42,5 cm
Weight: 4 kg