ContactUsLinkComponent
Electrolysis apparatus acc.Hoffmann
Electrolysis apparatus acc.Hoffmann
Catalog # BINH1002899
Supplier:  3B Scientific
undefined
Electrolysis apparatus acc.Hoffmann
Catalog # BINH1002899
Supplier:  3B Scientific
Specifications
  • Descrizione:
    Hofmann’s Voltameter, incl. gas collection tube, platinum electrodes, plastic hose with levelling bulb, stand ring, universal bosshead, stand baseplate with rod, dimensions ∼800×150 mm, rod Ø×H 12 mm×750 mm, retaining plate 120×110 mm
  • Conf:
    1