Isofreeze racks with temperature indicator
Isofreeze racks, PP, yellow-green, with temperature indicator, 96, 141×99×44 mm
Nome del fornitore: DELTALAB
Vai a
Product Details & Documents
PP, with clear lid.
- Filled with a non toxic gel that maintains 4 °C temperature for almost four hours when the rack is removed from the freezer
- A colour indicator begins to change from green to yellow as the temperature rises above 0 degrees
Documents
Certificates
Cerca per numero di lottoSpecifiche
- N. di fori:96
- Descrizione:Isofreeze racks, PP, yellow-green, with temperature indicator
- L×P×A:141×99×44 mm