Brushes, swivel-head, for flasks
Catalog # 149-1210
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Specifications
- Handle material:Legno
- Descrizione:Brush with wooden handle, swivel-head
- Lunghezza totale:420 mm
- Conf:1
Specifications
About this item
For intensive and gentle cleaning. Fits flasks from 500 to 5000 ml with SGJ/NS 29 openings.