Order Entry
Switzerland
ContactUsLinkComponent
N(α)-Acetyl-L-tryptophan
N(α)-Acetyl-L-tryptophan
Catalog # BACE4003876.0025
Supplier:  BACHEM BIOCHEMICA
CAS Number:  
N(α)-Acetyl-L-tryptophan
Catalog # BACE4003876.0025
Supplier:  BACHEM BIOCHEMICA
CAS Number:  

Specifications

  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :246.3 g/mol
EINECS:214-935-9
CAS Number:1218-34-4
Formula:C₁₃H₁₄N₂O₃
MDL Number:MFCD00065976