Solid phase extraction cartridges, HyperSep™ Diol
A short alkyl chains with polar functional groups are ideal for the isolation of polar compounds.
- Normal phase extraction of polar compounds
- Purification of polar compounds
HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customisable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.
Packaging: HyperSep-96 are available either in packs of 100 individual wells or in pre-assembled plates.
A short alkyl chains with polar functional groups are ideal for the isolation of polar compounds.
- Normal phase extraction of polar compounds
- Purification of polar compounds
HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customisable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.
Packaging: HyperSep-96 are available either in packs of 100 individual wells or in pre-assembled plates.
Product Family Options
- Formato della colonnaFaseDimensioni particelleCapacitàPeso letto
- CartucciaDiol40 - 60 µm1 ml50 mg1 * 100 ST447.00 SFr.
- CartucciaDiol40 - 60 µm1 ml100 mg1 * 100 ST455.00 SFr.
- CartucciaDiol40 - 60 µm15 ml2000 mg1 * 20 ST381.00 SFr.
- CartucciaDiol40 - 60 µm25 ml5000 mg1 * 20 ST419.00 SFr.
- CartucciaDiol40 - 60 µm75 ml10000 mg1 * 10 ST465.00 SFr.
96 well plates and individual wells
- DescrizioneRiempimento
- 96-well plate, HyperSep™ Diol10 mg1 * 1 ST678.00 SFr.