Certified vial kits
Supplier: THERMO NATIONAL SCIENTIFIC
Certificates
About this item
These certified vials are made of clear Type 1, 33 expansion borosilicate glass or Type 2, 51 expansion borosilicate glass.
- Fully lot-tested including HPLC and GC analysis
- Available with or without ID patch
- Bonded caps prevents leaking
- Fixed micro vials or inserts for limited volume sampling
Delivery: Certificate of conformance included in every pack.
Frequently Bought Together