Eyewash bottle
Eyewash bottle, empty, 1000 ml
Supplier: DELTALAB
Jump to:
Product Details & Documents
Made of low density polyethylene, this eyewash bottle is flexible and easy to squeeze.
- Has an eye cup with snap-on cap
- Eyecup has a perforated plate to break up the stream providing a gentle spray to the eye
- Capped air vent valve prevents drawback of contaminated eye wash into the bottle
- Contaminated solution exits through a drain tube, which rotates in any direction
- Instructions for use printed in various languages
Documents
Certificates
Search by Lot NumberSpecifications
- Descrizione:Eyewash bottle, empty, 1000 ml