Orders LinkContactUsLinkComponent
Microtubes racks | Avantor
Microtubes racks
Microtubes racks
Supplier:  DELTALAB
Certificates
Microtubes racks
5 Options Available Below
Product Details & Documents
Space saving racks, made of PP, designed to accommodate microtubes for freezer storage.
  • Alpha-numeric matrix allows accurate sample identification
  • Lateral handles allow easy handling of the racks
  • Use with 1,5/2,0 ml microtubes and 2 ml cryotubes
  • Suitable for freezer use down to −80 °C
  • Autoclavable
Documents
Product Family Options
Product Information
  • Colore
    N. di fori
    per Ø provette
    L×P×A
    Availability
    Price
  • blu
    80 (5×16)
    11 mm
    225×65×27 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 74.00
    SFr. 74.00
    1 * 5 ST
       SFr. 74.00
  • verde
    80 (5×16)
    11 mm
    225×65×27 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 74.00
    SFr. 74.00
    1 * 5 ST
       SFr. 74.00
  • naturale
    80 (5×16)
    11 mm
    225×65×27 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 69.80
    SFr. 69.80
    1 * 5 ST
       SFr. 69.80
  • rosso
    80 (5×16)
    11 mm
    225×65×27 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 74.00
    SFr. 74.00
    1 * 5 ST
       SFr. 74.00
  • giallo
    80 (5×16)
    11 mm
    225×65×27 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 74.00
    SFr. 74.00
    1 * 5 ST
       SFr. 74.00
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.