Surgical scissors, single use, Aesculap® SUSI®
Straight, sharp/blunt, 145 mm
Nome del fornitore: Aesculap
Surgical scissors, single use, Aesculap® SUSI®
Codice catalogo AESCBC324SU
Nome del fornitore: Aesculap
Vai a
Product Details & Documents
These disposable scissors are made from a high performance polymer. The special composition of this high-grade polymer ensures an outstanding quality and provides optimal mechanical properties.
- Single use reduces risk of cross-contamination
- Reduction of processing costs in hospitals
- Sterilised in a validated process
The material ensures a safe, easy and environmentally friendly removal through incineration in the regular hospital waste removal system.
Confezione: The scissors are sterile packed in single peel packs. The single peel packs are presented in dispenser boxes containing an indicated quantity of units as described in this leaflet.
Documents
Specifiche
- Tipo:Straight, sharp/blunt
- Lunghezza:145 mm