Heating Cords
Nome del fornitore: ISOPAD
Vai a
Informazioni su questo articolo
Flexible heating cords consist of insulated glass silk, flexible woven fabric with interior heating conductors.
- Nominal temperature: 450 °C
- Frequency: 50 to 60 Hz
6 varianti
Product Details & Documents
Flexible heating cords consist of insulated glass silk, flexible woven fabric with interior heating conductors.
- Nominal temperature: 450 °C
- Frequency: 50 to 60 Hz
Product Family Options
Informazioni sul prodotto
- LunghezzaDisponibilitàPrice
- ISOP175192-000Heating cords, 50 WSpecifiche:
More
Specifications
Cat. No.ISOP175192-000 - ISOP209402-000Heating cords, 160 WSpecifiche:
More
Specifications
Cat. No.ISOP209402-000 - ISOP490504-000Heating cords, 220 WSpecifiche:
More
Specifications
Cat. No.ISOP490504-000 - ISOP030246-000Heating cords, 330 WSpecifiche:
More
Specifications
Cat. No.ISOP030246-000 - ISOP920078-000Heating cords, 490 WSpecifiche:
More
Specifications
Cat. No.ISOP920078-000 - ISOP927312-000Heating cords, 710 WSpecifiche:
More
Specifications
Cat. No.ISOP927312-000