ContactUsLinkComponent
Earmuffs, Peltor™ SportTac™
  111-2215
 :  3M
undefined
Earmuffs, Peltor™ SportTac™
  111-2215
 :  3M
  • Altezza ripiano:
    26 dB
  • Colore:
    nero/rosso
  • Descrizione:
    Earmuffs, Peltor™ SportTac™, with foldable headband
  • Conf:
    1
  • Peso:
    318 g
These earmuffs offer a smooth, level-dependent hearing protection function that electronically cuts off abrupt, loud sound. They rapidly detect loud impulse noise which is cut out, while offering pleasant sound reproduction without interference.
  • Lightweight (318 g), with collapsible headband for easy storage
  • Interchangeable shells makes it easy to change colours
  • Smooth active-volume function that helps prevent sound being cut off abruptly
  • Automatic shut off
  • Battery lifespan about 600 hours, two AAA batteries
  • Audio input for an external radio
Designed for hunting and sports shooting.
SNR: 26 dB