High visibility rain trousers, Essential Hi-Vis, Class 2
Nome del fornitore: FRISTADS KANSAS
Certificati
Informazioni su questo articolo
Waterproof trousers made of PU coated polyester jersey with 'stretch' features.
- Elasticated waist
- Belt loops
- Adjustable legs
Weight: 170 g/m²
Certifications: EN 471 (2-2), EN 343 (3-2)
Frequently Bought Together