Handheld refractometer, IFT
Industrial fluid tester, IFT 40, 0 - 30, 1,3330 - 1,3860 nD, 0,5
Catalog # 635-2110
Supplier: REICHERT
Jump to:
Product Details & Documents
This handheld refractometer is designed to determine ratio or concentration of water-soluble industrial fluids or lubricants.
Specifications
- Modello:IFT 40
- Range di misura:0 - 30
- Risoluzione:0,5
- Descrizione:Industrial fluid tester
- Seq:02
- Indice di rifrazione:1,3330 - 1,3860 nD
- Cat. No.:635-2110
- Bilance:0,5