HTS Caco-2 assay system, PET membrane, BioCoat™, Corning
Codice catalogo 734-1078
Nome del fornitore: Corning
Specifiche
- Descrizione:HTS Caco-2 assay system, inc. media and supplements, for 120 individual 3-day Caco-2 assays
- Seq:02
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:ISO 14001 - Bedford
- Conf:5
Specifiche
Informazioni su questo articolo
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 assay system, PET membrane contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat fibrillar collagen 24-well insert system.