Sample tubes
Marketsource
Nome del fornitore: BIOZYM
Vai a
Informazioni su questo articolo
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
With or without screw caps.
19 varianti
Product Details & Documents
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
With or without screw caps.
Product Family Options
Without screw caps
Informazioni sul prodotto
- CapacitàFondoColoreMaterialeTipoSterilitàDisponibilitàPrice
- BZYM710002without side labelSpecifiche:MarketsourceMarketsource
- BZYM710015with side labelSpecifiche:MarketsourceMarketsource
- BZYM710016with side labelSpecifiche:MarketsourceMarketsource
- BZYM710006with side labelSpecifiche:MarketsourceMarketsource
- BZYM710020with side labelSpecifiche:MarketsourceMarketsource
- BZYM710021with side labelSpecifiche:MarketsourceMarketsource
- BZYM710011with side labelSpecifiche:MarketsourceMarketsource
- BZYM710012with side labelSpecifiche:MarketsourceMarketsource
With screw caps
Informazioni sul prodotto
- CapacitàFondoColoreMaterialeTipoSterilitàDisponibilitàPrice
- BZYM710051without side labelwithout side label0,5 mlConical bottomtrasparentePPscrew cap without attachment loopSteriliLog in per disponibilità1 * 500 STSpecifiche:MarketsourceMarketsource
- BZYM710055without side labelwithout side label0,5 mlFree standingtrasparentePPscrew cap without attachment loopSteriliLog in per disponibilità1 * 500 STSpecifiche:MarketsourceMarketsource
- BZYM710061without side labelwithout side label0,5 mlConical bottomtrasparentePPscrew cap with attachment loopSteriliLog in per disponibilità1 * 500 STSpecifiche:MarketsourceMarketsource
- BZYM710065without side labelwithout side label0,5 mlFree standingtrasparentePPscrew cap with attachment loopSteriliLog in per disponibilità1 * 500 STSpecifiche:MarketsourceMarketsource
- BZYM710052with side labelwith side label1,5 mlConical bottomtrasparentePPscrew cap without attachment loopSteriliLog in per disponibilità1 * 500 STSpecifiche:MarketsourceMarketsource
- BZYM710056with side labelwith side label1,5 mlFree standingtrasparentePPscrew cap without attachment loopSteriliLog in per disponibilità1 * 500 STSpecifiche:MarketsourceMarketsource
- BZYM710062with side labelwith side label1,5 mlConical bottomtrasparentePPscrew cap with attachment loopSteriliLog in per disponibilità1 * 500 STSpecifiche:MarketsourceMarketsource
- BZYM710066with side labelwith side label1,5 mlFree standingtrasparentePPscrew cap with attachment loopSteriliLog in per disponibilità1 * 500 STSpecifiche:MarketsourceMarketsource
- BZYM710053with side labelwith side label2,0 mlConical bottomtrasparentePPscrew cap without attachment loopSteriliLog in per disponibilità1 * 500 STSpecifiche:MarketsourceMarketsource
- BZYM710063with side labelwith side label2,0 mlConical bottomtrasparentePPscrew cap with attachment loopSteriliLog in per disponibilità1 * 500 STSpecifiche:MarketsourceMarketsource
- BZYM710067with side label, graduated in 0,25 ml divisionswith side label, graduated in 0,25 ml divisions2,0 mlFree standingtrasparentePPscrew cap with attachment loopSteriliLog in per disponibilità1 * 500 STSpecifiche:MarketsourceMarketsource
Screw caps
Informazioni sul prodotto
- Codice catalogoDescrizionePer provetteColoreDisponibilitàPrice
- Screw cap with inserted ring, EPR0,5/1,5/2,0 mltrasparenteLog in per disponibilità1 * 500 STSFr. 0.00