Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 SFr.
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cannucce criogeniche
Storage cane for cryogenic vials, Cryovial®
Storage cane for cryogenic vials, Cryovial®
Storage cane for cryogenic vials, Cryovial®
BZYM710550
:
BIOZYM
Storage cane for cryogenic vials, Cryovial®
BZYM710550
:
BIOZYM
Descrizione:
Aluminium stand for up to 5 tubes of sizes 1 and 2 ml
Conf:
12