Shell jacket, GEN-Y Cocona®
Nome del fornitore: FRISTADS KANSAS
Informazioni su questo articolo
Breathable jacket made of 100% polyester with 3,5-layer Cocona® PU laminate fabric. The fabric is windproof and waterproof.
- Dot-reinforcement at shoulders, front and lower sleeves
- Fleece-lined collar, outer and inside storm flap over front zip
- Double front pockets with logo zip, two inside pockets
- Raglan sleeves, reflector piping at hem
- Detachable hood with snap fasteners
- Adjustable strap with reflector tape on sleeves
Weight: 119 g/m²
21 varianti
Product Details & Documents
Breathable jacket made of 100% polyester with 3,5-layer Cocona® PU laminate fabric. The fabric is windproof and waterproof.
- Dot-reinforcement at shoulders, front and lower sleeves
- Fleece-lined collar, outer and inside storm flap over front zip
- Double front pockets with logo zip, two inside pockets
- Raglan sleeves, reflector piping at hem
- Detachable hood with snap fasteners
- Adjustable strap with reflector tape on sleeves
Weight: 119 g/m²
Documents
Specifications for Product Family
Specifiche
- Catalog No:
- KWIN100963331XS
- KWIN100963331S
- KWIN100963331M
- KWIN100963331L
- KWIN100963331XL
- KWIN1009633312XL
- KWIN1009633313XL
- KWIN100963530XS
- KWIN100963530S
- KWIN100963530M
- KWIN100963530L
- KWIN100963530XL
- KWIN1009635302XL
- KWIN1009635303XL
- KWIN100963940XS
- KWIN100963940S
- KWIN100963940M
- KWIN100963940L
- KWIN100963940XL
- KWIN1009639402XL
- KWIN1009639403XL
- Modello:
- Gen-Y
- Gen-Y
- Gen-Y
- Gen-Y
- Gen-Y
- Gen-Y
- Gen-Y
- Gen-Y
- Gen-Y
- Gen-Y
- Gen-Y
- Gen-Y
- Gen-Y
- Gen-Y
- Gen-Y
- Gen-Y
- Gen-Y
- Gen-Y
- Gen-Y
- Gen-Y
- Gen-Y
- Applicazioni per abiti:
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Genere:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Tipo di chiusura:
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Tessuto:
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Peso:
- 119 g/m²
- 119 g/m²
- 119 g/m²
- 119 g/m²
- 119 g/m²
- 119 g/m²
- 119 g/m²
- 119 g/m²
- 119 g/m²
- 119 g/m²
- 119 g/m²
- 119 g/m²
- 119 g/m²
- 119 g/m²
- 119 g/m²
- 119 g/m²
- 119 g/m²
- 119 g/m²
- 119 g/m²
- 119 g/m²
- 119 g/m²
Specifiche
Product Family Options
Informazioni sul prodotto
- ColoreTaglia dell'abbigliamentoVestibilità del capoDisponibilitàPrice
- Specifiche:
More
Specifications
Cat. No.KWIN100963331XSModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN100963331SModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN100963331MModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN100963331LModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN100963331XLModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN1009633312XLModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN1009633313XLModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN100963530XSModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN100963530SModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN100963530MModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN100963530LModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN100963530XLModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN1009635302XLModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN1009635303XLModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN100963940XSModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN100963940SModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN100963940MModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN100963940LModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN100963940XLModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN1009639402XLModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m² - Specifiche:
More
Specifications
Cat. No.KWIN1009639403XLModelloGen-YApplicazioni per abitiOutdoorRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyesterPeso119 g/m²