High visibility rain jacket, Essential, class 3
Basic rain jacket made of polyurethane-coated nylon jersey with 'stretch' function.
- Detachable drawstring hood
- Buttons and zip
- Adjustable cuffs
Weight: 170 g/m²
Certifications: EN 471 (3-2), EN 343 (3-2)
Basic rain jacket made of polyurethane-coated nylon jersey with 'stretch' function.
- Detachable drawstring hood
- Buttons and zip
- Adjustable cuffs
Weight: 170 g/m²
Certifications: EN 471 (3-2), EN 343 (3-2)
Documents
- Catalog No:
- KWIN1010121302XS
- KWIN1010121301XS
- KWIN101012130S
- KWIN114041-171M
- KWIN114041-171L
- KWIN1010121301XL
- KWIN1010121302XL
- KWIN1010121303XL
- KWIN1010122302XS
- KWIN1010122301XS
- KWIN101012230S
- KWIN101012230M
- KWIN101012230L
- KWIN1010122301XL
- KWIN1010122302XL
- KWIN1010122303XL
- Modello:
- Class 3
- Class 3
- Class 3
- Class 3
- Class 3
- Class 3
- Class 3
- Class 3
- Class 3
- Class 3
- Class 3
- Class 3
- Class 3
- Class 3
- Class 3
- Class 3
- Applicazioni per abiti:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Genere:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Tipo di chiusura:
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Tessuto:
- Polyurethane-coated nylon
- Polyurethane-coated nylon
- Polyurethane-coated nylon
- Polyurethane-coated nylon
- Polyurethane-coated nylon
- Polyurethane-coated nylon
- Polyurethane-coated nylon
- Polyurethane-coated nylon
- Polyurethane-coated nylon
- Polyurethane-coated nylon
- Polyurethane-coated nylon
- Polyurethane-coated nylon
- Polyurethane-coated nylon
- Polyurethane-coated nylon
- Polyurethane-coated nylon
- Polyurethane-coated nylon
- Peso:
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
Product Family Options
- ColoreTaglia dell'abbigliamento
Specifications
Cat. No.KWIN1010121302XSModelloClass 3Applicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyurethane-coated nylonPeso170 g/m²Specifications
Cat. No.KWIN1010121301XSModelloClass 3Applicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyurethane-coated nylonPeso170 g/m²Specifications
Cat. No.KWIN101012130SModelloClass 3Applicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyurethane-coated nylonPeso170 g/m²Specifications
Cat. No.KWIN114041-171MModelloClass 3Applicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyurethane-coated nylonPeso170 g/m²Specifications
Cat. No.KWIN114041-171LModelloClass 3Applicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyurethane-coated nylonPeso170 g/m²Specifications
Cat. No.KWIN1010121301XLModelloClass 3Applicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyurethane-coated nylonPeso170 g/m²Specifications
Cat. No.KWIN1010121302XLModelloClass 3Applicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyurethane-coated nylonPeso170 g/m²Specifications
Cat. No.KWIN1010121303XLModelloClass 3Applicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyurethane-coated nylonPeso170 g/m²Specifications
Cat. No.KWIN1010122302XSModelloClass 3Applicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyurethane-coated nylonPeso170 g/m²Specifications
Cat. No.KWIN1010122301XSModelloClass 3Applicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyurethane-coated nylonPeso170 g/m²Specifications
Cat. No.KWIN101012230SModelloClass 3Applicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyurethane-coated nylonPeso170 g/m²Specifications
Cat. No.KWIN101012230MModelloClass 3Applicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyurethane-coated nylonPeso170 g/m²Specifications
Cat. No.KWIN101012230LModelloClass 3Applicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyurethane-coated nylonPeso170 g/m²Specifications
Cat. No.KWIN1010122301XLModelloClass 3Applicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyurethane-coated nylonPeso170 g/m²Specifications
Cat. No.KWIN1010122302XLModelloClass 3Applicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyurethane-coated nylonPeso170 g/m²Specifications
Cat. No.KWIN1010122303XLModelloClass 3Applicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenereUnisexSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessutoPolyurethane-coated nylonPeso170 g/m²