Pipette tips, PreCision Line
Marketsource
Nome del fornitore: BIOZYM
Vai a
Informazioni su questo articolo
Transparent, yellow or blue; sterile or non sterile.
- Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
- Autoclavable
- Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
Certifications: DIN 12650
18 varianti
Product Details & Documents
Transparent, yellow or blue; sterile or non sterile.
- Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
- Autoclavable
- Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
Certifications: DIN 12650
Product Family Options
For Eppendorf, Brand and Socorex pipettes
Informazioni sul prodotto
- Stile puntaVolumeSteriliPer uso conConfezioneDisponibilitàPrice
- Crystal10 µl-10 µl Eppendorf, Brand and Socorex pipettes10 bags of 1000 tipsLog in per disponibilità1 * 10 STSpecifiche:Marketsource
More
Specifications
Cat. No.BZYM760001Marketsource - Crystal10 µl-10 µl Eppendorf, Brand and Socorex pipettes50 bags of 1000 tipsLog in per disponibilità1 * 50 STSpecifiche:Marketsource
More
Specifications
Cat. No.BZYM760003Marketsource - Crystal10 µl-10 µl Eppendorf, Brand and Socorex pipettes10 racks of 96 tipsLog in per disponibilità1 * 10 STSpecifiche:Marketsource
More
Specifications
Cat. No.BZYM760005Marketsource - Crystal10 µl+10 µl Eppendorf, Brand and Socorex pipettes10 racks of 96 tipsLog in per disponibilità1 * 10 STSpecifiche:Marketsource
More
Specifications
Cat. No.BZYM760008Marketsource
For Biozym PreCision, Biohit Proline, Gilson P-2 und P-10, Finn Digital, Rainin Pipet-Classic PR2 und PR10, Rainin Pipet-Lite SL-2 and SL-10 pipettes
Informazioni sul prodotto
- Stile puntaVolumeSteriliPer uso conConfezioneDisponibilitàPrice
- Standard10 µl-10 µl Sartorius Proline, Gilson P-2/P-10, Rainin Pipet-Classic PR2/PR10 + Pipet-Lite SL-2/SL-1050 bags of 1000 tipsLog in per disponibilità1 * 50 STSpecifiche:Marketsource
More
Specifications
Cat. No.BZYM760013Marketsource
For all popular pipettes
Informazioni sul prodotto
- Stile puntaVolumeSteriliPer uso conConfezioneDisponibilitàPrice
- Specifiche:Marketsource
More
Specifications
Cat. No.BZYM760201Marketsource - Specifiche:Marketsource
More
Specifications
Cat. No.BZYM760203Marketsource - Specifiche:Marketsource
More
Specifications
Cat. No.BZYM760205Marketsource - Specifiche:Marketsource
More
Specifications
Cat. No.BZYM760211Marketsource - Specifiche:Marketsource
More
Specifications
Cat. No.BZYM760213Marketsource - Specifiche:Marketsource
More
Specifications
Cat. No.BZYM760215Marketsource - Specifiche:Marketsource
More
Specifications
Cat. No.BZYM760218Marketsource - Specifiche:Marketsource
More
Specifications
Cat. No.BZYM761001Marketsource - Specifiche:Marketsource
More
Specifications
Cat. No.BZYM761003Marketsource - Specifiche:Marketsource
More
Specifications
Cat. No.BZYM761005Marketsource - Specifiche:Marketsource
More
Specifications
Cat. No.BZYM761011Marketsource - Specifiche:Marketsource
More
Specifications
Cat. No.BZYM761013Marketsource - Specifiche:Marketsource
More
Specifications
Cat. No.BZYM761018Marketsource