VWR®, Super Optimal Broth (SOB) Broth, per biotecnologie
Catalog # J906-500G
Supplier: VWR Chemicals
CAS Number:
Specifications
- Tipo di conf.:Bottiglia di plastica per i solidi
- Conf:500 g
- Descrizione:SOB broth
Specifications
About this item
Utilizzato per la preparazione terreno di crescita e trasformazione di cellule competenti E. coli
SOB broth growth medium is nutritionally rich, containing casein hydrolysate and yeast extract as sources of nitrogenous compounds and growth factors, plus magnesium sulphate as a supply of magnesium ions necessary for DNA replication.
When dissolved at 25,5 g/L of water, yields tryptone (20 g/L), yeast extract (five g/L), sodium chloride (0,5 g/L).
Store SOB broth powder at room temperature (18 to 26 °C). Store prepared medium cold (2 to 8 °C).
Caution: Note: Medium should be autoclaved or sterile-filtered. Allow the solution to cool to 50 °C or below before adding supplemental components and antibiotic. Add 20 ml of One M MgSO₄.