Reinforcement Patches, kwintet
Catalog # KWIN100388940
Supplier: FRISTADS KANSAS
Specifications
- Colore:nero
- Descrizione:Reinforcement patches for knee, thighs and elbows, pack of two
- Seq:01
- Conf:1
Specifications
About this item
Reinforcement patches for e.g. knees, thighs and elbows. Made of 72% polyamide, 24% Kevlar® and 4% elastane